missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethyl-3-heptanol, erythro + threo, 98+%, Thermo Scientific™
492.58 DKK - 1808.45 DKK
Chemical Identifiers
CAS | 19549-74-7 |
---|---|
Molecular Formula | C9H20O |
Molecular Weight (g/mol) | 144.26 |
MDL Number | MFCD00046585 |
InChI Key | NOSOEGQGQOMHBF-UHFFFAOYNA-N |
Synonym | 3,5-dimethyl-3-heptanol, 3-heptanol, 3,5-dimethyl, 3,5-dimethyl-3-heptanol, erythro + threo, acmc-20ance, 4-heptanol,3,5-dimethyl, ethyl methyl 2-methylbutyl carbinol |
PubChem CID | 140546 |
IUPAC Name | 3,5-dimethylheptan-3-ol |
SMILES | CCC(C)CC(C)(O)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499926
|
Thermo Scientific Alfa Aesar
B20896.09 |
10 g |
492.58 DKK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409936
|
Thermo Scientific Alfa Aesar
B20896.18 |
50 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used in the synthesis of tertiary 14C-labelled nonylphenol isomers..
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
19549-74-7 | |
144.26 | |
NOSOEGQGQOMHBF-UHFFFAOYNA-N | |
140546 | |
CCC(C)CC(C)(O)CC |
C9H20O | |
MFCD00046585 | |
3,5-dimethyl-3-heptanol, 3-heptanol, 3,5-dimethyl, 3,5-dimethyl-3-heptanol, erythro + threo, acmc-20ance, 4-heptanol,3,5-dimethyl, ethyl methyl 2-methylbutyl carbinol | |
3,5-dimethylheptan-3-ol |
Specifications
19549-74-7 | |
MFCD00046585 | |
NOSOEGQGQOMHBF-UHFFFAOYNA-N | |
3,5-dimethylheptan-3-ol | |
140546 | |
≥98% |
C9H20O | |
3,5-dimethyl-3-heptanol, 3-heptanol, 3,5-dimethyl, 3,5-dimethyl-3-heptanol, erythro + threo, acmc-20ance, 4-heptanol,3,5-dimethyl, ethyl methyl 2-methylbutyl carbinol | |
CCC(C)CC(C)(O)CC | |
144.26 | |
144.25 | |
3,5-Dimethyl-3-heptanol, erythro + threo |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures