Learn More
Butenafine Hydrochloride, MedChemExpress
MedChemExpress Butenafine Hydrochloride (KP363 Hydrochloride) is a synthetic benzylamine antifungal, works by inhibiting the synthesis of sterols by inhibiting squalene epoxidase.
Brand: Medchem Express HY-17396
Description
Butenafine Hydrochloride (KP363 Hydrochloride) is a synthetic benzylamine antifungal, works by inhibiting the synthesis of sterols by inhibiting squalene epoxidase.Specifications
Butenafine Hydrochloride | |
101827-46-7 | |
5 g | |
C23H28ClN | |
KP363 Hydrochloride | |
CN(CC1=CC=C(C(C)(C)C)C=C1)CC2=C3C=CC=CC3=CC=C2.[H]Cl | |
353.93 | |
Research |
Solid | |
White | |
Research | |
COVID-19-immunoregulation | |
DMSO : 25 mg/mL (70.64 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.83 mM; Need ultrasonic) | |
353.93 | |
99.96% |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only