Learn More
Macherey-Nagel™ Chromabond™ ec Polypropylene Column
SPE columns for vacuum manifold extraction
Brand:  Macherey-Nagel™ 730015ASP
 View more versions of this product
Product Code. 12891061
  • 4750.00 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Automated systems for CHROMABOND™ vacuum manifolds
  • Polypropylene column, polyethylene filter element
  • High loading capacity and separation efficiency
  • Constant back pressure, airtight sealing
  • Standard Luer port For automated solid phase extraction, parallel flash purification/synthesis, high throughput screening/purification.
Specifications
Specifications
Silica
SPE
Solid Phase Extraction
Solid Phase Extraction Column
6 mL
1000 mg
Chromabond C18 ec
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Chromabond™ ec Polypropylene Column > Column Capacity: 6mL; Adsorbent Weight: 1000mg