Learn More
DUPA(OtBu)-OH, MedChemExpress
MedChemExpress DUPA(OtBu)-OH is a DUPA precursor. DUPA is used as the targeting moiety to actively deliver Docetaxel (DTX) for treatment of Prostate-Specific Membrane Antigen (PSMA) expressing prostate cancer.
Brand:  Medchem Express HY-103591
Encompass_Preferred
 View more versions of this product
Product Code. 16433868
  • 2396.95 DKK / 10mg
Explore more special offers
Description
Description
DUPA(OtBu)-OH is a DUPA precursor. DUPA is used as the targeting moiety to actively deliver Docetaxel (DTX) for treatment of Prostate-Specific Membrane Antigen (PSMA) expressing prostate cancer.
Specifications
Specifications
DUPA(OtBu)-OH
1026987-94-9
10 mg
C23H40N2O9
DMSO : 125 mg/mL (255.85 mM; Need ultrasonic)
488.57
98.0%
Solid
White
Research
Cancer-programmed cell death
O=C(OC(C)(C)C)CC[C@@H](C(OC(C)(C)C)=O)NC(N[C@H](C(OC(C)(C)C)=O)CCC(O)=O)=O
488.57
Research
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
DUPA(OtBu)-OH, MedChemExpress