Learn More
DUPA(OtBu)-OH, MedChemExpress
MedChemExpress DUPA(OtBu)-OH is a DUPA precursor. DUPA is used as the targeting moiety to actively deliver Docetaxel (DTX) for treatment of Prostate-Specific Membrane Antigen (PSMA) expressing prostate cancer.
Brand: Medchem Express HY-103591
Description
DUPA(OtBu)-OH is a DUPA precursor. DUPA is used as the targeting moiety to actively deliver Docetaxel (DTX) for treatment of Prostate-Specific Membrane Antigen (PSMA) expressing prostate cancer.Specifications
DUPA(OtBu)-OH | |
1026987-94-9 | |
50 mg | |
C23H40N2O9 | |
DMSO : 125 mg/mL (255.85 mM; Need ultrasonic) | |
488.57 | |
98.0% |
Solid | |
White | |
Research | |
Cancer-programmed cell death | |
O=C(OC(C)(C)C)CC[C@@H](C(OC(C)(C)C)=O)NC(N[C@H](C(OC(C)(C)C)=O)CCC(O)=O)=O | |
488.57 | |
Research |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only