Learn More
GSK2018682, MedChemExpress
MedChemExpress GSK2018682 is an agonist for S1P1 and S1P5 receptor with pEC50s of 7.7 and 7.2, respectively, and has no agonist activity towards human S1P2, S1P3, or S1P4. GSK2018682 is used in the research of multiple sclerosis.
Brand: Medchem Express HY-19511
Description
GSK2018682 is an agonist for S1P1 and S1P5 receptor with pEC50s of 7.7 and 7.2, respectively, and has no agonist activity towards human S1P2, S1P3, or S1P4. GSK2018682 is used in the research of multiple sclerosis.Specifications
GSK2018682 | |
White | |
C22H21ClN4O4 | |
DMSO : ≥ 125 mg/mL (283.52 mM) | |
440.88 | |
98.25% | |
Solid |
1034688-30-6 | |
Research | |
100 mg | |
O=C(O)CCCN1C=CC2=C1C=CC=C2C3=NOC(C4=CC(Cl)=C(OC(C)C)N=C4)=N3 | |
440.88 | |
Research | |
COVID-19-immunoregulation |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only