missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Light Module Cold White 6,500 K

Light strips arranged on the side walls of the interior

Brand:  Memmert™ HPP260ecoT7

 View more versions of this product

Product Code. 17199794

  • 26430.00 DKK / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 10 strips for model 110
  • 14 strips for model 260/410/750
  • Programme-controlled dimming from 0 to 100 %(in 1 % steps)
  • Ramp programming in combination with temperature
  • Only with TwinDISPLAY; not in combination with entry ports F6, F7, D8
Specifications

Specifications

White
HPP260eco
Only with TwinDISPLAY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Memmert™ Light Module Cold White 6,500 K > HPP260eco

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.