Learn More
Usbeck Laborgeraete™ Nylon Universal Scissors
Brand:  Usbeck Laborgeraete™ 4281
 View more versions of this product
Product Code. 10075651
  • 105.91 DKK / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Specifications
Universal Scissors
230 mm
Nylon
Documents
One moment while we fetch your results.
Product Title
Usbeck Laborgeraete™ Nylon Universal Scissors > Length: 230mm