Learn More
Hellma™ Quartz Flow Through Cells Far UV
Brand:  Hellma™ 170-000-2-40
 View more versions of this product
Product Code. 12647201
  • 5985.00 DKK / Each
Estimated Shipment: 08-08-2025
Explore more special offers
Specifications
Flow-through Cell Far UV
Quartz
2 mm
Product Title
Hellma™ Quartz Flow Through Cells Far UV > Light Path: 2mm