Learn More
Tenovin-3, MedChemExpress
MedChemExpress Tenovin-3 is a p53 activator.
Brand: Medchem Express HY-19339
Description
Tenovin-3 is a p53 activator.Specifications
Tenovin-3 | |
1011301-27-1 | |
50 mg | |
C18H21N3OS | |
DMSO : ≥ 31 mg/mL (94.67 mM) | |
327.44 | |
99.86% |
Solid | |
White | |
Research | |
Cancer-programmed cell death | |
O=C(NC(NC1=CC=C(N)C=C1)=S)C2=CC=C(C(C)(C)C)C=C2 | |
327.44 | |
Research |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only